Learn More
S2 Media 15x100mm Cetrimide Agar, 10/pack
Supplier:  S2 Media 5060
Encompass_Preferred
Cetrimide Agar is widely recommended for use in the examination of cosmetics, clinical specimens, and for evaluating the efficacy of disinfectants against Pseudomonas aeruginosa. It is also used in the microbiological examination of nonsterile pharmaceutical products. P. aeruginosa is an environmental organism and an important nosocomial pathogen.• Product meets BAM, CLSI and Harmonized USP/EP/JP reference methods and performance criteria.• Cetrimide Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5906