Learn More
Uline 15 x 18"" 2 Mil Reclosable Bags, 500/cs
Supplier:  Uline S17749
Encompass_Preferred
Single-track zipper closure keeps out dirt and moisture. All virgin polyethylene film meets FDA and USDA requirements.
Catalog No. NC1539515