Learn More
Medchemexpress LLC 16:0-18:2 PA sodium | 322647-59-6 | MFCD00674300 | 95.0% | 694.89 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14706750MG
Encompass_Preferred
16:0-18:2 PA sodium is a glycerophospholipid that acts as a bioactive lipid and secondary messenger. It is intended for research use only and not for human, veterinary, or therapeutic applications. It is supplied as a solid and can be used as a biological marker to indicate pathological states.
  • Regulates various enzyme activities.
  • Functions as a bioactive lipid and secondary messenger.
  • Associated with hydrogen peroxide production in neutrophils.
  • Stimulates protein kinase action for neutrophil respiratory burst.
  • Participates in Ras mediated signaling pathways.
  • Supplied in powder form.
  • Color is white to off-white.
Catalog No. 50-003-59909