Learn More
Medchemexpress LLC (±)16(17)-DiHDPA | 1345275-27-5 | 99.0 % | 50 UG
SDP
Supplier:  Medchemexpress LLC HY12044250UG
Encompass_Preferred
(±)16(17)-DiHDPA is a racemic epoxygenase metabolite of docosahexaenoic acid (DHA) supplied for research use. It has been reported to inhibit platelet aggregation at concentrations below those affecting thromboxane synthesis.
  • Racemic epoxygenase metabolite of docosahexaenoic acid.
  • Inhibits platelet aggregation at concentrations below those affecting thromboxane synthesis.
  • Purity 99.0% and molecular weight 362.50 g/mol.
  • Available in 50 UG and 100 UG research quantities.
Catalog No. 50-002-33526