Learn More
Medchemexpress LLC 16α,17α-Epoxyprogesterone | 1097-51-4 | 50 G
SDP
Supplier:  Medchemexpress LLC HYN399450G
Encompass_Preferred
16α,17α-Epoxyprogesterone is an important steroid, serving as an intermediate for many hormonal drugs such as Hydrocortisone and Megestrol. It can further biotransform into another significant intermediate, 11α-hydroxy-16α,17α-epoxyprogesterone, to manufacture additional compounds.
  • Serves as an intermediate for hormonal drugs
  • Can biotransform into other intermediates
Catalog No. 50-004-18869