Learn More
AdipoGen 16-epi-Latrunculin B (100 µg)
SDP
Supplier:  AdipoGen AGCN20034C100
Encompass_Preferred
Chemical. CAS: 444911-05-1. Formula: C20H29NO5S. MW: 395.5. Isolated from marine sponge Negombata magnifica. Antiviral (against herpes simplex type 1 virus (HSV-1)). Cytotoxic. Depolymerizes actin filaments (F-actin).
Catalog No. 50-205-3381