Learn More
Medchemexpress LLC 16α-hydroxyprednisolone | 13951-70-7 | MFCD00872019 | 99.8% | 376.44 g·mol⁻1 | C21H28O6 | 50 G

Supplier: Medchemexpress LLC HY11758050G
16α-Hydroxyprednisolone is a high-purity steroid metabolite provided as a solid reference standard for research use, commonly employed in studies of glucocorticoid metabolism and CYP3A-mediated biotransformation.
- High purity (99.78%) suitable for analytical reference and in vitro studies.
- Solid form for ease of storage and handling.
- Molecular formula C21H28O6 and molecular weight 376.44 g·mol⁻1.
- Available in a 50 g pack for bulk research applications.
- Intended for research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.