Learn More
Sino Biological Recombinant Human RAB27B Protein (His Tag)
Supplier:  Sino Biological 16007H07H100
Encompass_Preferred
A DNA sequence encoding the human RAB27B (NP_004154.2) (Thr2-Lys215) was expressed with an N-terminal polyhistidine tag.
Catalog No. 50-161-9780