Learn More
Sino Biological Recombinant Human Calcitonin receptor Protein (ECD, hFc Tag)
Supplier:  Sino Biological 16023H02H100
Encompass_Preferred
A DNA sequence encoding the human CALCR (NP_001733.1) (Met1-Ile153) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9816