Learn More
Sino Biological Recombinant Human CLSTN1 Protein (ECD, His Tag)
Supplier:  Sino Biological 16035H08H50
Encompass_Preferred
A DNA sequence encoding the human CLSTN1 (NP_001009566.1) (Met1-Thr859) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9831