Learn More
Sino Biological Recombinant Human ILT7/LILRA4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 16058H08H50
Encompass_Preferred
A DNA sequence encoding the human LILRA4 (P59901.2) (Met1-Asn446) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9861