Learn More
LAB RESEARCH PRODUCTS 16 cu ft Pharmacy Refrigerator & Auto-Defrost Freezer Combo, 13 cu ft refrigerator / 3 cu ft freezer, double solid swing doors, R600a refrigerant, NEMA 5-15 plug, 31 in W x 35.5 in D x 76 in H
Supplier:  LAB RESEARCH PRODUCTS PHABTHCRFC16A
Encompass_Preferred
This 16 cu ft pharmacy refrigerator and auto-defrost freezer combination provides reliable temperature control and sample security for clinical and medical environments. It pairs an energy-efficient variable-speed compressor with a natural hydrocarbon refrigerant and digital microprocessor controls to maintain uniform temperatures, enable rapid recovery after door openings, and protect vaccines and critical samples.
  • 16 cu ft total storage (13cu ft refrigerator, 3cu ft freezer)
  • Adjustable refrigerator range 1°C to 10°C and freezer range -35°C to -15°C
  • Individual microprocessor controllers w/digital displays and °C/°F switchable settings
  • Audible and visual high/low temperature, power-loss, and door alarms.
  • 12V battery backup for controller, alarms, monitoring, and event logging
  • Variable speed compressor with 1300-4000 rpm range
  • Ext dimensions 31 in W x 35.5 in D x 76 in H; product weight 262 lbs.
  • Catalog No. NC3343808
    Only null left
    Add to Cart
    Request a Quote