Learn More
Educational Innovations 16OZ SODIUM POLYACRYLATE
SDP
Supplier:  Educational Innovations GB6B
Encompass
This fine white powder instantly turns liquid water into a slush-like solid substance. Technically speaking, the polymer absorbs from 500 to 1,000 times its own weight in water.
Catalog No. NC9010821