Learn More
Avanti Polar Lipids 17:0-17:1-17:0 D5 TG, 958760-74-2, MFCD22416552, powder 1mg
Supplier:  Avanti Polar Lipids 860903P1MG
Encompass
Molecular Formula C54H97D5O6, Formula Weight 852.415, Storage Temperature -20°C, 1,3(d5)-diheptadecanoyl-2-(10Z-heptadecenoyl)-glycerol
Catalog No. NC1634955