Learn More
Medchemexpress LLC 17-azido-3,6,9,12,15-pentaoxaheptadecyl 4-methylbenzenesulfonate | 906007-10-1 | MFCD29918254 | 98.6% | C19H31N3O8S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14035310MG
Encompass_Preferred
Azide-PEG6-Tos is an azide-functionalized PEG6 tosylate linker for bioconjugation and PROTAC synthesis. It supplies an azide reactive handle and a tosyl leaving group for downstream functionalization, facilitating click chemistry and linker assembly in small-molecule and bioconjugate research.
  • High purity of 98.6% suitable for synthetic applications.
  • Azide functional group enables copper-catalyzed and strain-promoted click reactions.
  • Tosylate leaving group allows facile nucleophilic substitution and modification.
  • PEG6 spacer provides flexibility and improves solubility in polar solvents.
  • Liquid form, colorless to light yellow, for easy handling and dosing.
  • Stable when stored under recommended conditions to maintain integrity.
  • Available in small milligram sizes for discovery and linker development.
Catalog No. 50-002-61452