Learn More
Medchemexpress LLC 17-dimethylaminoethylamino-17-demethoxygeldanamycin | 467214-20-6 | MFCD08457919 | 99.5% | 616.75 g·mol⁻¹ | C32H48N4O8 | 100 MG

Supplier: Medchemexpress LLC HY10389100MG
Alvespimycin (17-DMAG) is a geldanamycin-derived small-molecule inhibitor of heat shock protein 90 (Hsp90). It binds Hsp90 with an EC50 of approximately 62 nM and is supplied as a high-purity solid for research use in preclinical and biochemical studies.
- Potent Hsp90 inhibitor with reported EC50 ≈ 62 nM.
- High purity (99.52% by HPLC).
- Molecular weight 616.75 g·mol⁻¹; formula C32H48N4O8.
- Physical appearance: pale purple to purple solid.
- Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months).
- Available in milligram pack sizes and DMSO solutions for ready reconstitution.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.