Learn More
Cayman Chemical 17-EstradIol-d3 10mg
SDP
Supplier:  Cayman Chemical 3897410 MG
Encompass_Preferred
An internal standard for the quantification of 17β-estradiol by GC- or LC-MS
Catalog No. 50-291-8773