Learn More
Medchemexpress LLC 17β-HSD1-IN-1 | 2760791-96-4 | 99.8% | 335.40 g/mol | C21H21NO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1454335MG
Encompass_Preferred
17β-HSD1-IN-1 is a selective small-molecule inhibitor of 17β-hydroxysteroid dehydrogenase type 1, intended for laboratory research. It inhibits 17β-HSD1 with an IC50 of 5.6 nM and shows substantially lower activity against 17β-HSD2 (IC50 3,155 nM). The compound is a solid, white to off-white powder (C21H21NO3, MW 335.40 g/mol) supplied at high reported purity for in vitro and preclinical studies.
  • Selective 17β-HSD1 inhibition (IC50 5.6 nM).
  • Low off-target activity against 17β-HSD2 (IC50 3,155 nM).
  • High reported purity suitable for research (99.79%).
  • Available as a solid powder for solution preparation or formulation.
  • Designed for in vitro and preclinical investigation of estrogen-related pathways.
Catalog No. 50-002-21611