Learn More
Medchemexpress LLC 17-Hydroxyneomatrine | 2306139-04-6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN82285MG
Encompass_Preferred
17-Hydroxyneomatrine, extracted from Sophora flavescens, effectively inhibits the growth of human cervical carcinoma Hela cells. It possesses a broad range of biological activities.
  • Inhibits growth of human cervical carcinoma Hela cells
  • Antibacterial activity
  • Anti-allergy activity
  • Anti-tumor activity
  • Anti-arrhythmia activity
  • Swelling-subsiding diuresis
  • Immunizing functions
  • Biological regulation functions
Catalog No. 50-004-19697