Learn More
Medchemexpress LLC 17α-Hydroxyprogesterone (standard) | 68-96-2 | 99.8% | 50 MG

Supplier: Medchemexpress LLC HYB0891R50MG
17α-Hydroxyprogesterone (Standard) is the analytical standard of 17α-Hydroxyprogesterone. This product is intended for research and analytical applications. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
- Application: Used as a reference standard in assays for qualitative, quantitative, and methodological research experiments, particularly in techniques like HPLC, GC, and MS.
- Molecular weight: 330.47
- Formula: C₂₁H₃₀O₃
- Appearance: Solid, white to off-white.
- Structure classification: Steroids.
- Initial source: Endogenous metabolite; disease markers for endocrine diseases and nervous system disorders.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.