Learn More
Cayman Chemical 17-OctadecynoIc AcId 10mg
SDP
Supplier:  Cayman Chemical 9027010 MG
Encompass_Preferred
A fatty acid alkyne; a suicide inhibitor of leukotriene B4 20-hydroxylase and renal CYP450 ω-hydroxylase; completely inhibits the bradykinin-dependent transport of sodium chloride in rat TALH cells; has been used as a click chemistry probe for labeling palmitoylation substrates in vitro; has been used to study the formation of lipid droplets in live THP-1 macrophages and C. elegans
Catalog No. 50-289-4860