Learn More
Enzo Life Sciences 17-Octadecynoic acid (10mg). CAS: 34450-18-5
SDP
Supplier:  Enzo Life Sciences BMLEI1030010
Encompass_Preferred
Potent suicide inhibitor of LTB4 ω-oxidase (IC50<5µM). Potent and selective inhibitor of fatty acid ω-hydroxylase (IC50<100 nM). Blocks biosynthesis of 20-HETE. Purity: ≥98% (HPLC). Solubility: Soluble in ethanol (25mg/ml) or DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0546