Learn More
Medchemexpress LLC 17-Octadecynoic acid | 34450-18-5 | 98.0% | 280.45 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY101016
Encompass_Preferred
17-ODYA is a potent inhibitor of CYP450 ω-hydroxylase, specifically hindering the formation of 20-hydroxyeicosatetraenoic acid (20-HETE), epoxyeicosatrienoic acids, and dihydroxyeicosatrienoic acids. It has demonstrated the ability to counteract isoproterenol-induced apoptosis and necrosis in cultured cardiomyocytes. Additionally, it functions as a click chemistry reagent due to its alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules.
  • Inhibits ω-hydroxylase activity.
  • Reduces formation of eicosanoids.
  • Attenuates cell death in cardiomyocytes.
  • Functions as a click chemistry reagent.
  • Can be used for chemical reactions with azide groups.
  • Has shown effects on renal function in rats.
  • May offer cardioprotective benefits.
Catalog No. 50-003-64422