Learn More
Medchemexpress LLC 17-Phenyl trinor PGF 10mg | 130209-76-6 | 430.6 g/mol | C26H38O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY116161A10MG
Encompass_Preferred
17-Phenyl trinor PGF2α isopropyl ester is a synthetic prostaglandin F2α analogue used as a research reagent for prostaglandin receptor and endocrinology studies (CAS 130209-76-6). It is provided in small milligram quantities for analytical and biological experiments and is typically supplied with characterization data.
  • Synthetic prostaglandin F2α analogue suitable for receptor studies.
  • High purity, commonly reported as ≥98% by some suppliers.
  • Available in small milligram pack sizes for research applications.
  • Soluble in common organic solvents; consult supplier documentation for solvent guidance.
  • Useful as a reference standard for pharmacology and assay validation.
Catalog No. 50-002-29793