Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 17-Phenyl trinor PGF 50mg | 130209-76-6 | 430.6 g/mol | C26H38O5 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY116161A50MG

Encompass_Preferred

17-phenyl trinor prostaglandin F2α isopropyl ester (CAS 130209-76-6) is a synthetic F-series prostaglandin analog and FP receptor agonist used in biomedical research. It is supplied as a small, premeasured solid for laboratory studies of prostaglandin signaling and ocular hypotensive activity.

  • Prostaglandin F2α analog with activity at FP receptors.
  • Suitable for in vitro and in vivo research applications.
  • Available as a 50 MG premeasured solid for laboratory use.
  • CAS 130209-76-6 provided for unambiguous identification.
  • Molecular formula C26H38O5; molecular weight 430.6 g/mol.

Catalog No. 50-002-29791


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.