Learn More
Cayman Chemical 17 R-ResolvIn D1 100ug
SDP
Supplier:  Cayman Chemical 13060100 UG
Encompass_Preferred
An aspirin-triggered epimer of RvD1; reduces human leukocyte transendothelial migration (EC50 = ~30 nM), and limits leukocyte infiltration in a mouse model of peritonitis
Catalog No. 50-295-3100