Learn More
Cayman Chemical 17(R)-Resolvin D1, 528583-91-7, 25ug
SDP
Supplier:  Cayman Chemical 1306025
Encompass_Preferred
Molecular Formula C22H32O5, Purity ≥95%, Formula Weight 376.5, Synonyms: 7S,8R,17R-trihydroxy-4Z,9E,11E,13Z,15E19Z-docosahexaenoic acid, Aspirin-triggered Resolvin D1, 17-epi-Resolvin D1, AT-RvD1, 17(R)-RvD1; An aspirin-triggered epimer of RvD1; reduces human leukocyte transendothelial migration (EC50 = ~30 nM), and limits leukocyte infiltration in a mouse model of peritonitis.
Catalog No. NC0353892
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.