Learn More
Cayman Chemical 17 R-ResolvIn D3 10UG
SDP
Supplier:  Cayman Chemical 900288010 UG
Encompass_Preferred
An aspirin-triggered epimer of resolvin D3; has been found in mouse inflammatory exudates; reduces transmigration of neutrophils into the peritoneal cavity, as well as decreases the levels of IL-6 and increases the levels of IL-10 in inflammatory exudate in a mouse model of zymosan-induced peritonitis at 10 ng/animal; activates GPR32 in a β-arrestin reporter assay and increases phagocytosis to a greater degree in CHO cells overexpressing GPR32 compared to mock-transfected cells; increases phagocytosis of etoposide-generated tumor cell debris by monocyte-derived macrophages in H460 human lung carcinoma cells in a concentration-dependent manner; inhibits tumor growth in a mouse model of Lewis lung carcinoma at 0.6 UG/kg per day
Catalog No. 50-289-5732