Learn More
Cayman Chemical 17 S-HDHA-d5 100ug
SDP
Supplier:  Cayman Chemical 29797100 UG
Encompass_Preferred
An internal standard for the quantification of 17(S)-HDHA by GC- or LC-MS
Catalog No. 50-298-9044