Learn More
Cayman Chemical 17 SHETE Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 1001130550 UG
Encompass_Preferred
A potent inhibitor of proximal tubule ATPase activity; is a CYP450 metabolite of arachidonic acid; inhibits proximal tubule ATPase activity by 70% at 2 µM
Catalog No. 50-289-0666