Learn More
Ace Glass, Inc. 17 liter graduated cylindrical jar with side indents
SDP
Supplier:  Ace Glass, Inc. 623317
Encompass
17L graduated cylindrical jar with side indents for easier handling. Made of heavy wall glass. Also available with poly safety coating. Not for use with heat, pressure, or vacuum applications.
Catalog No. 50-972-773