Learn More
Cayman Chemical 17oxo7 Z 10 Z 13 Z 15 E 100UG
SDP
Supplier:  Cayman Chemical 9000347100 UG
Encompass_Preferred
A metabolite of lipoxygenase-mediated oxidation of DPA; activates Nrf2-dependent antioxidant gene expression, acts as a PPARγ agonist (EC50 = ~200 nM), and inhibits pro-inflammatory cytokine and nitric oxide production at biological concentration ranges (5-25 µM)
Catalog No. 50-289-4712