Learn More
Cayman Chemical 17phenl trInor ProstglndI 10mg
SDP
Supplier:  Cayman Chemical 1682110 MG
Encompass_Preferred
A derivative of 17-phenyl trinor PGF2α ethyl amide; the amide of 17-phenyl trinor PGF2α amide may be hydrolyzed to the free acid form in human eyes; like other amide isomers of F-type prostaglandins, 17-phenyl trinor PGF2α may have similar FP receptor activation and IOP reduction activities; has a lower hydrolysis rate in primary human corneas compared to isopropyl ester-protected prodrugs
Catalog No. 50-296-6201