Learn More
Cayman Chemical 17phenyl trinor13 14dihydro P
SDP
Supplier:  Cayman Chemical 102901 MG
Encompass_Preferred
17-phenyl trinor-13,14-dihydro PGA2 is a synthetic PG analog whose biological activity has not been widely reported. The PGF2α analog latanoprost, which bears the same lower side chain features, has been approved as a pharmaceutical for the treatment of glaucoma.{5338}
Catalog No. 50-289-0208