Learn More
Avanti Polar Lipids 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (ammonium salt), 474922-90-2, MFCD02262251, 25 mg
Supplier:  Avanti Polar Lipids 880130C25MG
Encompass
Formula Weight: 2801.465, Purity >99%, Storage Temperature: -20°C.
Catalog No. NC1252063