Learn More
Medchemexpress LLC 18α-Glycyrrhetinic acid | 1449-05-4 | 470.68 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN037550MG
Encompass_Preferred
18α-Glycyrrhetinic acid is a diet-derived compound that functions as an inhibitor of NF-κB and an activator of proteasome. It acts as a pro-longevity and anti-aggregation factor in multicellular organisms and also induces apoptosis.
  • Diet-derived compound
  • Inhibitor of NF-κB
  • Activator of proteasome
  • Serves as a pro-longevity factor
  • Acts as an anti-aggregation factor
  • Induces apoptosis
Catalog No. 50-003-88864