Learn More
Medchemexpress LLC 18α-Glycyrrhetinic acid | 1449-05-4 | MFCD00064897 | 470.68 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0375
Encompass_Preferred
18α-Glycyrrhetinic acid is a diet-derived compound that acts as an inhibitor of NF-κB and an activator of proteasome. It serves as a pro-longevity and anti-aggregation factor in multicellular organisms and induces apoptosis.
  • Reduces LX-2 cell numbers
  • Increases percentage of LX-2 cells in phase G0/G1 and decreases in phase S
  • Increases apoptosis in LX-2 cells
  • Induces expression of PPAR-γ and alters certain cell cycle and apoptosis-related proteins
  • Inhibits NF-κB DNA-binding activity
  • Enhances life span of *C. elegans* strains
  • Delays paralysis in *C. elegans*
  • Reduces Aβ deposits in *C. elegans*
Catalog No. 50-003-89598