Learn More
Hampton Research HR4-945/Mounted CryoLoop - 20 micron, 18 x 0.05-0.1 mm - 25 pack
SDP
Supplier:  Hampton Research HR4945
Encompass
HR4-945/Mounted CryoLoop - 20 micron, 18 x 0.05-0.1 mm - 25 pack. Please note items from this supplier are non-returnable.
Catalog No. NC0375426