Learn More
Pacer Global LLC 182cm2 Tissue Culture Flask, Treated, Vent Cap, Sterile, 5 per pack, 40 per case
SDP
Supplier:  Pacer Global LLC MLSCFTSV182J/DEL
Encompass_Preferred
182cm2 Tissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachment. Vent caps feature 0.22µm hydrophobic membrane. Flask is printed with lot number and expiration date for easy identification and tracking. Flask is made from Polystyrene and is RNase/DNase free. Cap is made from Polyethylene. 5/pack, 8 packs/case. Additional info on MLS-CFTSV-182J: Temperature Range: -20°~+50°. Non-pyrogenic. Bulk packed in zip-lock bag. Not Autoclavable. Working volume: 400 ml. Recommended working volume: 125 ml. Growth area: 182 cm. Shelf Life: 3 years after month of production. Sterilized by gamma irradiation SAL10-6 (ISO11137). Manufactured in a Class 100,000 cleanroom environment. Manufactured under ISO13485:2016 and ISO9001:2015 quality management system.
Catalog No. NC1957345