Learn More
Uline Jumbo Nylon Cable Ties, Natural, 100
Supplier:  Uline S11144
Encompass_Preferred
Super-tough nylon ties secure heavy duct wires and cables, 18 cable bundles up to 5 diameter, UL Listed 66 nylon, Locks tightly - Wont stretch or slide, Temperature range -40 to 180 F
Catalog No. NC2671249