Learn More
Chemglass Life Sciences 19/22 Inner Joint, Cut Drip, on 17mm OD x 1.8mm
SDP
Supplier:  Chemglass Life Sciences CG11704
Encompass_Preferred
Full or medium length standard taper inner joint having a drip tip. Joints are available uncut or with the drip tip being cut at a 45º angle. Tooled on the same tubing sizes as the CG-100 inner joints. 19/22 Inner Joint, Cut Drip, on 17mm OD x 1.8mm.
Catalog No. CG11704