Learn More
Medchemexpress LLC 19-Norlanost-5-en-11-one, 3,24,25-trihydroxy-9-methyl-, (3β,9β,10α)- | 88930-16-9 | 5 MG

Supplier: Medchemexpress LLC HYN43125MG
Bryodulcosigenin is an extract of the roots of Bryonia dioica with anti-inflammatory effect. It is for research use only.
- Purity: 99.21%
- Appearance: Solid, white to off-white
- Structure classification: Terpenoids, triterpenes
- Initial source: Plants
- Biological activity: Anti-inflammatory effect
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.