Learn More
Cayman Chemical 19R 20S-EpDPA 100ug
SDP
Supplier:  Cayman Chemical 28628100 UG
Encompass_Preferred
A DHA metabolite; prevents calcium-induced increases in NRCM spontaneous beating at 30 nM
Catalog No. 50-294-1067