Learn More
eMolecules​ 59084-16-1 | 1-Acetylpiperidine-4-carbonyl chloride | Combi-Blocks | MFCD02094017 | 189.640 | C8H12ClNO2 | 95.000 | CC(=O)N1CCC(CC1)C(Cl)=O | 25g | 335354881
Supplier:  eMolecules​ 335354881
Encompass_Preferred
1-Acetylpiperidine-4-carbonyl chloride | Combi-Blocks | 59084-16-1 | MFCD02094017 | 189.640 | C8H12ClNO2 | 95.000 | CC(=O)N1CCC(CC1)C(Cl)=O | 25g | 335354881
Catalog No. NC3059981