Learn More
Cayman Chemical 1ArachIdonyld5racglycero 100ug
SDP
Supplier:  Cayman Chemical 362152100 UG
Encompass_Preferred
An internal standard for the quantification of 1-AG by GC- or LC-MS
Catalog No. 50-291-1684