Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1beta,11beta,12alpha,15beta)-1,11,12,14,15-pentahydroxy-13,20-epoxypicras-3-ene-2,16-dione | 21499-66-1 | MFCD00221753 | 99.5% | 410.42 g·mol⁻¹ | C20H26O9 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN301410MG

Encompass_Preferred

Bruceine D is a quassinoid natural product used as a research reagent that induces apoptosis and inhibits Notch signaling in cancer cell models. It is supplied as a high-purity solid suitable for in vitro biochemical and cell-based studies.

  • High purity: 99.5%.
  • Molecular formula: C20H26O9.
  • Molecular weight: 410.42 g·mol⁻¹.
  • Appearance: white to off-white solid.
  • Recommended solvent: DMSO; 100 mg/mL, ultrasonic assistance may be required.
  • Storage: store at 4°C protected from light; in solution store at -80°C up to 6 months or -20°C up to 1 month.
  • Common applications: in vitro apoptosis assays and cell-based mechanism-of-action studies.

Catalog No. 50-002-89503


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.