Learn More
Medchemexpress LLC 1beta,25-dihydroxycholecalciferol | 66791-71-7 | MFCD22124486 | 98.0% | 416.64 g/mol | C27H44O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY132925MG
Encompass_Preferred
Impurity B of Calcitriol (1β,25-dihydroxycholecalciferol) is an EP-specified impurity standard for analytical characterization of calcitriol. It is supplied as a high-purity reference material intended for method development, impurity identification, and quantitative analysis in pharmaceutical and biochemical laboratories.
  • High purity suitable for analytical reference.
  • Available in small, controlled package sizes for method development.
  • Molecular formula and weight provided for analytical calculations.
  • Suitable as a reference standard for HPLC and mass spectrometry.
  • Supplied with documentation for traceability and quality control.
Catalog No. 50-002-63052