Learn More
Climet Instruments Climet Instruments 1 CFM Zero Count Filter, Non-Autoclavable, ISO Class 3 Certified
SDP
Supplier:  Climet Instruments 01206504R
Encompass
The CLIMET INSTRUMENTS 1 CFM/50 LPM Zero Count Filter ensures a particle-free baseline essential for precise calibration and verification of Climet particle counters. Designed for critical environmental monitoring, this non-autoclavable filter meets stringent ISO Class 3 certification standards. Its unmatched reliability supports cleanroom compliance and preserves data integrity in scientific and regulated applications, making it ideal for users seeking accurate and dependable performance in controlled environments.
Catalog No. NC1673715