Learn More
Medchemexpress LLC 1E7-03 | 1565845-92-2 | 95.1% | 503.55 | C28H29N3O6 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1246441MG
Encompass_Preferred
1E7-03 is a low molecular weight tetrahydroquinoline derivative that targets protein phosphatase-1 (PP1) and has been reported to inhibit HIV-1 transcription in peer-reviewed studies. It is supplied as a research reagent with available documentation including a datasheet, certificate of analysis, and safety data sheet, and has specified storage and handling recommendations for long-term stability.
  • Low molecular weight tetrahydroquinoline derivative.
  • Targets protein phosphatase-1 (PP1).
  • Reported to inhibit HIV-1 transcription in published studies.
  • Supplied as a small-quantity research reagent (mg scale).
  • Purity approximately 95.1% as supplied.
  • Documented storage conditions for powder and solution stability.
  • Manufacturer provides datasheet, COA, and SDS.
Catalog No. 50-002-37150