Learn More
Berlin Packaging Corporation 1GAL CAN OBLONG W/ SEAL&CAP
Supplier:  Berlin Packaging Corporation 317164
Encompass
1 Gallon Silver Oblong Tin Metal Can With Seal And Continuous Thread Cap. Certified To Carry Liquid Hazardous Material In Packing Group Ii. Classified As Closed Head Steel Drum.
Catalog No. NC0709414
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.